Learn More
Bio IVT M DH GUINP CRY HEP 5 M CLLSV
Catalog No. 502310848
Encompass
Hepatocyte, Cryopreserved; Celsis In Vitro Technologies; From male Dunkin-Hartley guinea pig, pooled; Storage temperature:-150 deg.C; Used to study phase I and phase II metabolism; Size: 5 million cells per vial
Catalog No. 50-231-0848 Supplier Bio IVT Supplier No. M00105
Hepatocyte, Cryopreserved; Celsis In Vitro Technologies; From male Dunkin-Hartley guinea pig, pooled; Storage temperature:-150 deg.C; Used to study phase I and phase II metabolism; Size: 5 million cells per vial