Learn More
Zymo Research Corporation M-Digestion Buffer (2X), 15 ml
SDP
Catalog No. NC0881266
Encompass_Preferred
M-Digestion Buffer-2X (15 ml). For use with the EZ DNA Methylation™-Direct bisulfite conversion kits.
Catalog No. NC0881266 Supplier Zymo Research Corporation Supplier No. D50219
M-Digestion Buffer-2X (15 ml). For use with the EZ DNA Methylation™-Direct bisulfite conversion kits.