Learn More
TWD Scientific LLC Medium Polypropylene Weighing Funnel. Pack of 50.
SDP
Catalog No. 501173372 Shop All TWD Scientific LLC Products
Encompass_Preferred
Disposable polypropylene weighing funnel, size medium. Fits volumetric flask volumes (mL) of 100, 200, 250. Funnels have a maximum capacity of 10mL Solid/7mL Liquid. 50/Pack
Catalog No. 50-117-3372 Supplier TWD Scientific LLC Supplier No. DPWFPP2MPK
Disposable polypropylene weighing funnel, size medium. Fits volumetric flask volumes (mL) of 100, 200, 250. Funnels have a maximum capacity of 10mL Solid/7mL Liquid. 50/Pack