Learn More
Zymo Research Corporation M-Dissolving Buffer, 1.2 ml
SDP
Catalog No. 501251521
Encompass_Preferred
The M-Dissolving buffer facilitates the dissolution of the CT Conversion Reagent from a solid mixture into a liquid reagent. Highlights: Sold separately or as part of the EZ DNA Methylation-Gold Kits. Storage temperature: room temperature.
Catalog No. 50-125-1521 Supplier Zymo Research Corporation Supplier No. D50066
The M-Dissolving buffer facilitates the dissolution of the CT Conversion Reagent from a solid mixture into a liquid reagent. Highlights: Sold separately or as part of the EZ DNA Methylation-Gold Kits. Storage temperature: room temperature.