Learn More
Zymo Research Corporation M-Dissolving Buffer (500 μl)
SDP
Catalog No. 501251518
Encompass_Preferred
The M-Dissolving buffer facilitates the dissolution of the CT Conversion Reagent from a solid mixture into a liquid reagent. Highlights: Sold separately or as part of the EZ DNA Methylation-Gold Kits. Storage temperature: room temperature.
Catalog No. 50-125-1518 Supplier Zymo Research Corporation Supplier No. D50056
The M-Dissolving buffer facilitates the dissolution of the CT Conversion Reagent from a solid mixture into a liquid reagent. Highlights: Sold separately or as part of the EZ DNA Methylation-Gold Kits. Storage temperature: room temperature.