Learn More
Zymo Research Corporation M-Elution Buffer, 40 ml
SDP
Catalog No. 502037127
Encompass_Preferred
Used for eluting and storing bisulfite converted DNA in the EZ DNA Methylation, EZ DNA Methylation - Gold, EZ DNA Methylation - Direct kits, and EZ DNA Methylation - Lightning kits.
Catalog No. 50-203-7127 Supplier Zymo Research Corporation Supplier No. D50416
Used for eluting and storing bisulfite converted DNA in the EZ DNA Methylation, EZ DNA Methylation - Gold, EZ DNA Methylation - Direct kits, and EZ DNA Methylation - Lightning kits.