Learn More
MOLTOX Molecular Toxicology Inc m Endo Agar LES Plate, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator/Dark
SDP
Catalog No. 50948691
Encompass_Preferred
m Endo Agar, LES is used for enumerating coliforms in water by membrane filtration technique. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-691 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S80
m Endo Agar, LES is used for enumerating coliforms in water by membrane filtration technique. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.