Learn More
Hach Company m-Endo Broth, 100 mL glass bottle, 50 tests
Supplier:  Hach Company 2373542
Encompass
m-Endo broth detects total coliform bacteria (colonies with a greenish-gold sheen). For potable water samples, confirm with Lauryl Tryptose and Brilliant Green Bile Broth tubes. Samples that test positive for total coliform bacteria should be further analyzed for the presence of fecal coliform or E. coli using EC Medium or EC Medium with MUG respectively.
Catalog No. 50-199-5885