Learn More
Hach Company m-Endo Broth, Dehydrated 500 g
Supplier:  Hach Company 1462300
Encompass
500 g dehydrated m-Endo Broth is used for enumerating coliforms by the membrane filtration method.
Catalog No. 50-199-5090