Learn More
Lifeline Cell Technology VascuLife VEGF Endothelial Medium Complete Kit
SDP
Catalog No. 50311892
Encompass_Preferred
Includes VascuLife Low Serum Basal Medium 475 mL, VascuLife VEGF LifeFactors Kit (with Vascular Endothelial Growth Factor). Optimized for proliferation of human endothelial cells - Umbilical Vein Endothelial, aortic, other large-vessel.
Catalog No. 50-311-892 Supplier Lifeline Cell Technology Supplier No. LL0003
Includes VascuLife Low Serum Basal Medium 475 mL, VascuLife VEGF LifeFactors Kit (with Vascular Endothelial Growth Factor). Optimized for proliferation of human endothelial cells - Umbilical Vein Endothelial, aortic, other large-vessel.