Learn More
Lifeline Cell Technology M ENDO HU LIFELINE KT
SDP
Catalog No. 50311891
Encompass_Preferred
Includes VascuLife Low Serum Basal Medium 475 mL, VascuLife EnGS LifeFactors Kit (with Endothelial Cell Growth Supplement). Optimized for the culture of human endothelial cells - Human Umbilical Vein, aortic, other large-vessel.
Catalog No. 50-311-891 Supplier Lifeline Cell Technology Supplier No. LL0002
Includes VascuLife Low Serum Basal Medium 475 mL, VascuLife EnGS LifeFactors Kit (with Endothelial Cell Growth Supplement). Optimized for the culture of human endothelial cells - Human Umbilical Vein, aortic, other large-vessel.