Learn More
Alkali Scientific m Endo LES Agar 20mL Monoplate, 10/Pk
SDP
Catalog No. 5000662007
Encompass_Preferred
m Endo LES Agar, 20mL Monoplate, 10/Pk is a selective and differential culture medium designed for the detection and enumeration of coliform bacteria, particularly Escherichia coli, in water samples using the membrane filtration method. The formulation contains lactose, sodium lauryl sulfate, and sodium deoxycholate, which inhibit Gram positive organisms while allowing Gram negative coliforms to grow and produce characteristic red colonies with a metallic sheen. Each plate is pre poured with 20mL of sterile medium in a standard 100 x 15mm monoplate format, ensuring even membrane placement and consistent incubation conditions. This medium is widely used for drinking water testing, wastewater analysis, and environmental monitoring, and aligns with EPA, APHA, and Standard Methods protocols for microbiological water quality analysis.
Catalog No. 50-006-62007 Supplier Alkali Scientific Supplier No. LESP00
m Endo LES Agar, 20mL Monoplate, 10/Pk is a selective and differential culture medium designed for the detection and enumeration of coliform bacteria, particularly Escherichia coli, in water samples using the membrane filtration method. The formulation contains lactose, sodium lauryl sulfate, and sodium deoxycholate, which inhibit Gram positive organisms while allowing Gram negative coliforms to grow and produce characteristic red colonies with a metallic sheen. Each plate is pre poured with 20mL of sterile medium in a standard 100 x 15mm monoplate format, ensuring even membrane placement and consistent incubation conditions. This medium is widely used for drinking water testing, wastewater analysis, and environmental monitoring, and aligns with EPA, APHA, and Standard Methods protocols for microbiological water quality analysis.