Learn More
MilliporeSigma M-ENDO PREPOURED CASSETTES
Catalog No. NC9622915
Encompass
Pre-filled Milliflex Cassettes, product name: m-Endo LES, target Microorganisms: Coliform organisms, for use with the Milliflex system, 48/Pk.
Catalog No. NC9622915 Supplier MilliporeSigma Supplier No. MXSMEND48
Pre-filled Milliflex Cassettes, product name: m-Endo LES, target Microorganisms: Coliform organisms, for use with the Milliflex system, 48/Pk.