Learn More
MOLTOX Molecular Toxicology Inc M-Enterococcus Agar, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator/Dark
SDP
Catalog No. 50948809
Encompass_Preferred
m-Enterococcus Agar, also known as m Azide Agar, is a selective medium used in membrane filtration or plate pour technique for isolating and enumerating enterococci in water. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-809 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S009
m-Enterococcus Agar, also known as m Azide Agar, is a selective medium used in membrane filtration or plate pour technique for isolating and enumerating enterococci in water. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.