Learn More
NEOGEN m-Enterococcus Agar, 500 g
SDP
Catalog No. 502015624
Encompass_Preferred
m-Enterococcus Agar is used for the selective isolation and enumeration of enterococci by membrane filtration in a laboratory setting. m-Enterococcus Agar is not intended for use in the diagnosis of disease or other conditions in humans.m-Enterococcus Agar was first described by Slanetz et al. for the enumeration of enterococci by the membrane filtration technique. In 1957, Slanetz and Bartley modified this medium by adding triphenyl tetrazolium chloride (TTC). Increased recovery and larger colonies were obtained by incubating the inoculated membranes on the agar surface instead of on pads saturated with liquid medium. The membrane filtration method is simple to perform, does not require confirmation, and permits a direct count of enterococci in 48 hours. m-Enterococcus Agar is also referred to as m-Azide Agar.For more information visit https://www.neogen.com/solutions/Microbiology/m-enterococcus-agar
Catalog No. 50-201-5624 Supplier NEOGEN Supplier No. 700003444
m-Enterococcus Agar is used for the selective isolation and enumeration of enterococci by membrane filtration in a laboratory setting. m-Enterococcus Agar is not intended for use in the diagnosis of disease or other conditions in humans.m-Enterococcus Agar was first described by Slanetz et al. for the enumeration of enterococci by the membrane filtration technique. In 1957, Slanetz and Bartley modified this medium by adding triphenyl tetrazolium chloride (TTC). Increased recovery and larger colonies were obtained by incubating the inoculated membranes on the agar surface instead of on pads saturated with liquid medium. The membrane filtration method is simple to perform, does not require confirmation, and permits a direct count of enterococci in 48 hours. m-Enterococcus Agar is also referred to as m-Azide Agar.For more information visit https://www.neogen.com/solutions/Microbiology/m-enterococcus-agar