Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aobchem M-Ethylacetophenone | ≥97% | CAS 22699-70-3 | C10H12O | 5g

Catalog No. 5000992286
Encompass_Preferred

M-Ethylacetophenone | ≥97% | CAS 22699-70-3 | C10H12O | 5g

Catalog No. 50-009-92286 Supplier Aobchem Supplier No. A2099435G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M-Ethylacetophenone | ≥97% | CAS 22699-70-3 | C10H12O | 5g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.