Learn More
Hach Company m-FC with Rosolic Acid, 2 mL plastic ampules, pk/50
Supplier:  Hach Company 2428550
Encompass
Hach m-FC with rosalic acid is used to detect and enumerate fecal coliforms in wastewater using the membrane filtration method.
Catalog No. 50-199-5964