Learn More
Lifeline Cell Technology M FIBS HU LIFELINE KT
SDP
Catalog No. 50311896
Encompass_Preferred
Includes FibroLife Basal Medium 480 mL, FibroLife S2 LifeFactors Kit. Optimized for low serum (2%) culture of fibroblasts at rates equal to or greater than comparable media supplemented with 2-10% FBS. No antimicrobials, no phenol red.
Catalog No. 50-311-896 Supplier Lifeline Cell Technology Supplier No. LL0011
Includes FibroLife Basal Medium 480 mL, FibroLife S2 LifeFactors Kit. Optimized for low serum (2%) culture of fibroblasts at rates equal to or greater than comparable media supplemented with 2-10% FBS. No antimicrobials, no phenol red.