Learn More
Lifeline Cell Technology M FIBS HU LIFELINE KT
SDP
Catalog No. 50311890
Encompass_Preferred
Includes FibroLife Basal Medium 480 mL, HLL Supplement, FibroLife LifeFactors Kit. Optimized for the serum-free culture of fibroblasts at rates equal to or greater than media supplemented with 2-10% FBS. No antimicrobials, no phenol red.
Catalog No. 50-311-890 Supplier Lifeline Cell Technology Supplier No. LL0001
Includes FibroLife Basal Medium 480 mL, HLL Supplement, FibroLife LifeFactors Kit. Optimized for the serum-free culture of fibroblasts at rates equal to or greater than media supplemented with 2-10% FBS. No antimicrobials, no phenol red.