Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ M-FIX™ Spray Fixative

Catalog No. M1039811000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100mL
1L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1039811000 1L
M1039810102 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1039811000 Supplier MilliporeSigma™ Supplier No. 1039811000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as fixing agent for cytodiagnosis

  • Fixative spray
  • Wet fixation of gynecological non-gynecological preparations processed in Papanicolaou staining
  • Contains polyglycol in alcoholic solution
  • Rapid, secure fixation
  • Enables stabilization of the material upon alcohol fixation
  • Protective film against desiccation damage to specimen material
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 25°C
Quantity 1L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.