Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Fluoronitrobenzene, MP Biomedicals

Catalog No. ICN205203 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205203 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205203 Supplier MP Biomedicals, Inc Supplier No. 0520520310

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 402-67-5
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.10
MDL Number MFCD00007196
InChI Key WMASLRCNNKMRFP-UHFFFAOYSA-N
Synonym 3-fluoronitrobenzene, m-fluoronitrobenzene, m-nitrofluorobenzene, benzene, 1-fluoro-3-nitro, 3-fluoro-1-nitrobenzene, 1-fluoro-3-nitro-benzene, 3-fluoro-nitrobenzene, 3-nitro-fluorobenzene, pubchem2286, 3-fluoro nitro benzene
PubChem CID 9823
IUPAC Name 1-fluoro-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(F)=C1

Specifications

Quantity 100 g
Formula Weight 141.1
Chemical Name or Material M-Fluoronitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.