Learn More
Bioworld M-Green Yeast and Fungi Broth, 500 g
Catalog No. 50254472
Encompass_Preferred
M-Green Yeast and Fungi BrothWorking concentration: 73.23g/LA selective medium used for the detection of yeast and fungi in beveragespH: 4.6Dehydrated Appearance: homogeneous powderPrepared Appearance: green to blue-green brothSoluble in waterComposition:Pancreatic Digest of Casein Peptic Digest of Animal Tissue Yeast Extract Dextrose Magnesium Sulfate Potassium Phosphate Clarase Thiamine Bromcresol Green
Catalog No. 50-254-472 Supplier Bioworld Supplier No. 306260021
M-Green Yeast and Fungi BrothWorking concentration: 73.23g/LA selective medium used for the detection of yeast and fungi in beveragespH: 4.6Dehydrated Appearance: homogeneous powderPrepared Appearance: green to blue-green brothSoluble in waterComposition:Pancreatic Digest of Casein Peptic Digest of Animal Tissue Yeast Extract Dextrose Magnesium Sulfate Potassium Phosphate Clarase Thiamine Bromcresol Green