Learn More
Alkali Scientific M Heterotrophic Plate Count Agar (MHPC) 20mL Plate, 10/Pk
SDP
Catalog No. 5000662257
Encompass_Preferred
M Heterotrophic Plate Count (m HPC) Agar is a nutrient rich culture medium formulated for the enumeration of heterotrophic bacteria in water samples using the membrane filtration method. Each plate contains approximately 20 mL of agar that supports growth of a wide range of microorganisms requiring organic carbon sources. The formulation includes peptones and gelatin that provide nitrogen and carbon nutrients, enabling recovery of stressed or chlorine injured bacteria that might not grow on less supportive media. Heterotrophic plate count testing is commonly used to evaluate overall microbiological quality of drinking water, recreational water, and treated wastewater. Elevated counts may indicate distribution system issues, ineffective disinfection, or microbial regrowth in storage systems.
Catalog No. 50-006-62257 Supplier Alkali Scientific Supplier No. MHPP00
M Heterotrophic Plate Count (m HPC) Agar is a nutrient rich culture medium formulated for the enumeration of heterotrophic bacteria in water samples using the membrane filtration method. Each plate contains approximately 20 mL of agar that supports growth of a wide range of microorganisms requiring organic carbon sources. The formulation includes peptones and gelatin that provide nitrogen and carbon nutrients, enabling recovery of stressed or chlorine injured bacteria that might not grow on less supportive media. Heterotrophic plate count testing is commonly used to evaluate overall microbiological quality of drinking water, recreational water, and treated wastewater. Elevated counts may indicate distribution system issues, ineffective disinfection, or microbial regrowth in storage systems.