Learn More
General Laboratory Products M-HPC Agar, 250 mL, 9/cs
Catalog No. NC9029248
Encompass_Preferred
M-HPC Agar - 250ml, Easy-pour narrow mouth glass bottle, 9 per case. Base for the Enumeration of Heterotrophic Organisms in Water by Membrane.
Catalog No. NC9029248 Supplier General Laboratory Products Supplier No. 250EZ7096250
M-HPC Agar - 250ml, Easy-pour narrow mouth glass bottle, 9 per case. Base for the Enumeration of Heterotrophic Organisms in Water by Membrane.