Learn More
Bio IVT M HUFREPLA HEP-24WL CLTRE PLA
Catalog No. 502310900
Encompass
Hepatocyte, Fresh plated; Celsis In Vitro Technologies; From male human; Biocoat plate; Collagen coated with collagen overlay; Storage temp.: 5% CO2, 37 deg.C incubator; Size: 24-well culture plate
Catalog No. 50-231-0900 Supplier Bio IVT Supplier No. M91539
Hepatocyte, Fresh plated; Celsis In Vitro Technologies; From male human; Biocoat plate; Collagen coated with collagen overlay; Storage temp.: 5% CO2, 37 deg.C incubator; Size: 24-well culture plate