Learn More
Santa Cruz Biotechnology m-IgG2a BP-HRP, 50 µg/0.5 ml
Catalog No. NC3785879
Encompass
Mouse IgG2a binding protein (m-IgG2a BP) conjugated to horseradish peroxidase (HRP) is a strongly recommended alternative to conventional goat/rabbit anti-mouse IgG secondary antibodies for Western Blotting (WB) and immunohistochemical (IHC) signal enhancement. Mouse IgG2a binding protein is a highly specific reagent that provides strong signal with minimal background and virtually complete elimination of lot to lot variation associated with conventionally generated secondary antibodies. Mouse IgG2a binding protein (m-IgG2a BP) is suitable for for binding to most, but not all mouse monoclonal IgG2a antibodies; not suitable for use with mouse monoclonal IgG1, IgG2b, IgG3, IgM, IgA or IgE antibodies. Not cross reactive with human or rat IgG antibodies.For Research Use Only. Not Intended for Diagnostic or Therapeutic Use.
Catalog No. NC3785879 Supplier Santa Cruz Biotechnology Supplier No. SC542731
Mouse IgG2a binding protein (m-IgG2a BP) conjugated to horseradish peroxidase (HRP) is a strongly recommended alternative to conventional goat/rabbit anti-mouse IgG secondary antibodies for Western Blotting (WB) and immunohistochemical (IHC) signal enhancement. Mouse IgG2a binding protein is a highly specific reagent that provides strong signal with minimal background and virtually complete elimination of lot to lot variation associated with conventionally generated secondary antibodies. Mouse IgG2a binding protein (m-IgG2a BP) is suitable for for binding to most, but not all mouse monoclonal IgG2a antibodies; not suitable for use with mouse monoclonal IgG1, IgG2b, IgG3, IgM, IgA or IgE antibodies. Not cross reactive with human or rat IgG antibodies.For Research Use Only. Not Intended for Diagnostic or Therapeutic Use.