Learn More
Innovive Inc M-innotube 175/case, Sheet-to-tube enrichment; 100% paper pulp enrichment sheets are uniquely designed to transform into a tube shape to encourage rodents' natural climbing and nesting instincts.
Catalog No. NC2830118
Encompass_Preferred
Sheet-to-tube enrichment; 100% paper pulp enrichment sheets are uniquely designed to transform into a tube shape to encourage rodents' natural climbing and nesting instincts.Keeps rodents active and helps reduce aggression.Can be combined with Innorichment™ sheets to offer more enrichment and complex, higher-scoring nests.Each sheet can be rolled into a locked tube shape to provide shelter and nesting material. For mouse caging.
Catalog No. NC2830118 Supplier Innovive Inc Supplier No. MINNOTUBE
Sheet-to-tube enrichment; 100% paper pulp enrichment sheets are uniquely designed to transform into a tube shape to encourage rodents' natural climbing and nesting instincts.Keeps rodents active and helps reduce aggression.Can be combined with Innorichment™ sheets to offer more enrichment and complex, higher-scoring nests.Each sheet can be rolled into a locked tube shape to provide shelter and nesting material. For mouse caging.