Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-Iodophenol | 626-02-8 | 220.01 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW00736410G

Encompass_Preferred

m-Iodophenol is a Protaetia brevitarsis attractant. This chemical compound appears yellow to brown, existing as a solid below 42°C and a liquid above 44°C.

  • Molecular formula: C6H5IO
  • Molecular weight: 220.01
  • High purity: 99.89%
  • Soluble in DMSO (100 mg/mL)
  • Also soluble in various in vivo protocols.

Catalog No. 50-004-60352


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.