Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-Iodophenol | 626-02-8 | 220.01 | 5 G
SDP

Catalog No. 5000460323
Encompass_Preferred

m-Iodophenol is a protaetia brevitarsis attractant. It is suitable for research use only.

  • Purity: 99.89%
  • Molecular weight: 220.01
  • Formula: C6H5IO
  • CAS No.: 626-02-8
  • Appearance: <42°C Solid, >44°C Liquid
  • Color: Yellow to brown

Catalog No. 50-004-60323 Supplier Medchemexpress LLC Supplier No. HYW0073645G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-Iodophenol is a protaetia brevitarsis attractant. It is suitable for research use only.

  • Purity: 99.89%
  • Molecular weight: 220.01
  • Formula: C6H5IO
  • CAS No.: 626-02-8
  • Appearance: <42°C Solid, >44°C Liquid
  • Color: Yellow to brown

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.