Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-Iodophenol | 626-02-8 | 220.01 | 50 G
SDP

Catalog No. 5000460436
Encompass_Preferred

m-Iodophenol is a chemical compound known as a *protaetia brevitarsis* attractant. This product offers specific characteristics for various laboratory applications.

  • Appears as a solid below 42°C and a liquid above 44°C.
  • Color ranges from yellow to brown.
  • Store at 4°C, protected from light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.

Catalog No. 50-004-60436 Supplier Medchemexpress LLC Supplier No. HYW00736450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-Iodophenol is a chemical compound known as a *protaetia brevitarsis* attractant. This product offers specific characteristics for various laboratory applications.

  • Appears as a solid below 42°C and a liquid above 44°C.
  • Color ranges from yellow to brown.
  • Store at 4°C, protected from light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.