Learn More
Cayman Chemical m-isobutyl Ibuprofen, Purity: ≥98%, CAS: 66622-47-7, 50 mg
SDP
Catalog No. 502942390
Encompass_Preferred
A potential impurity in commercial preparations of ibuprofen
Catalog No. 50-294-2390 Supplier Cayman Chemical Supplier No. 4012750 MG
A potential impurity in commercial preparations of ibuprofen