Learn More
Cayman Chemical m-isobutyl Ibuprofen, Purity: ≥98%, CAS: 66622-47-7, 5 mg
SDP
Catalog No. 502942500
Encompass_Preferred
A potential impurity in commercial preparations of ibuprofen
Catalog No. 50-294-2500 Supplier Cayman Chemical Supplier No. 401275 MG
A potential impurity in commercial preparations of ibuprofen