Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-isobutyl ibuprofen | 66622-47-7 | MFCD08275582 | >=98.0% | 206.28 g/mol | C13H18O2 | 5 MG
SDP

Catalog No. 5000262935
Encompass_Preferred

m-Isobutyl Ibuprofen (Compound IMP-A) is an impurity produced during the synthesis of ibuprofen and is supplied as a research-grade impurity standard for analytical and quality-control applications. The material is intended for laboratory and research use only and should be handled according to the certificate of analysis and institutional safety procedures.

  • Reference impurity for analytical and quality-control use.
  • Intended for research use only, not for human or veterinary use.
  • Available in small milligram pack sizes suitable for method development.
  • Molecular formula C13H18O2; molecular weight 206.28 g/mol.
  • Storage conditions provided in the certificate of analysis; follow laboratory safety guidance.

Catalog No. 50-002-62935 Supplier Medchemexpress LLC Supplier No. HY1313915MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-Isobutyl Ibuprofen (Compound IMP-A) is an impurity produced during the synthesis of ibuprofen and is supplied as a research-grade impurity standard for analytical and quality-control applications. The material is intended for laboratory and research use only and should be handled according to the certificate of analysis and institutional safety procedures.

  • Reference impurity for analytical and quality-control use.
  • Intended for research use only, not for human or veterinary use.
  • Available in small milligram pack sizes suitable for method development.
  • Molecular formula C13H18O2; molecular weight 206.28 g/mol.
  • Storage conditions provided in the certificate of analysis; follow laboratory safety guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.