Learn More
Medchemexpress LLC M-isobutyl ibuprofen | 66622-47-7 | MFCD08275582 | ≥98.0% | C13H18O2 | 10MG
SDP
Catalog No. 5000261985
Encompass_Preferred
m-Isobutyl ibuprofen is a synthetic impurity (Ibuprofen impurity A) encountered during the manufacture of ibuprofen. It is supplied for research and analytical purposes as a small-mass reference standard used in impurity profiling, method development, and quality control. CAS 66622-47-7; molecular formula C13H18O2.
  • Intended for research use and analytical reference.
  • Suitable as an impurity reference for HPLC and GC method development.
  • Available in small pack sizes for laboratory workflows.
  • Provided with data sheet and certificate of analysis for lot-specific details.
  • Store under recommended conditions; consult certificate of analysis for stability information.
Catalog No. 50-002-61985 Supplier Medchemexpress LLC Supplier No. HY13139110MG
m-Isobutyl ibuprofen is a synthetic impurity (Ibuprofen impurity A) encountered during the manufacture of ibuprofen. It is supplied for research and analytical purposes as a small-mass reference standard used in impurity profiling, method development, and quality control. CAS 66622-47-7; molecular formula C13H18O2.
  • Intended for research use and analytical reference.
  • Suitable as an impurity reference for HPLC and GC method development.
  • Available in small pack sizes for laboratory workflows.
  • Provided with data sheet and certificate of analysis for lot-specific details.
  • Store under recommended conditions; consult certificate of analysis for stability information.