Learn More
Lifeline Cell Technology M KERATOCYT HU LIFELINE KT
SDP
Catalog No. 50311902
Encompass_Preferred
DermaLife K Calcium Free is serum free medium optimized for the culture of human epidermal keratinocytes. Calcium Chloride has been omitted form the Basal Medium, is supplied as an individual Life Factor. No antimicrobials and no phenol red
Catalog No. 50-311-902 Supplier Lifeline Cell Technology Supplier No. LL0029
DermaLife K Calcium Free is serum free medium optimized for the culture of human epidermal keratinocytes. Calcium Chloride has been omitted form the Basal Medium, is supplied as an individual Life Factor. No antimicrobials and no phenol red