Learn More
Lifeline Cell Technology M KERATOCYT LIFELINE KT
SDP
Catalog No. 50311895
Encompass_Preferred
Includes DermaLife Serum-Free Basal Medium 485 mL, DermaLife K LifeFactors Kit. Optimized for the culture of human keratinocytes over a period of at least 15 population doublings. Contains no antimicrobials and no phenol red.
Catalog No. 50-311-895 Supplier Lifeline Cell Technology Supplier No. LL0007
Includes DermaLife Serum-Free Basal Medium 485 mL, DermaLife K LifeFactors Kit. Optimized for the culture of human keratinocytes over a period of at least 15 population doublings. Contains no antimicrobials and no phenol red.