Learn More
AVANTOR MASTERFLEX (VWR LLC) ADCF Male Luer to 1/4"" L Barb Adapter, PP, 25/pk
Catalog No. NC0823294
Encompass
ADCF Male Luer to 1/4" L Barb Adapter, Polypropylene, 25/pk, Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization. (not recommended for autoclaving).
Catalog No. NC0823294 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3080026
ADCF Male Luer to 1/4" L Barb Adapter, Polypropylene, 25/pk, Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization. (not recommended for autoclaving).