Learn More
Bio IVT M MARMOSET MON-Y HEP 5 M CLLSV
Catalog No. 502310860
Encompass
Hepatocyte, Cryopreserved; Celsis In Vitro Technologies; From male Marmoset monkey, pooled; Storage temperature:-150 deg.C; Used to study phase I and phase II metabolism; Size: 5 million cells per vial
Catalog No. 50-231-0860 Supplier Bio IVT Supplier No. M00325
Hepatocyte, Cryopreserved; Celsis In Vitro Technologies; From male Marmoset monkey, pooled; Storage temperature:-150 deg.C; Used to study phase I and phase II metabolism; Size: 5 million cells per vial