Learn More
Lifeline Cell Technology M MEL HU LIFELINE KT
SDP
Catalog No. 50311906
Encompass_Preferred
Includes BronchiaLife Basal Medium 485 mL, BronchiaLife SAE LifeFactors Kit. Optimized for the serum-free culture of human small airway epithelial cells. Supports the growth of these cells over a period of at least 15 population doublings.
Catalog No. 50-311-906 Supplier Lifeline Cell Technology Supplier No. LL0039
Includes BronchiaLife Basal Medium 485 mL, BronchiaLife SAE LifeFactors Kit. Optimized for the serum-free culture of human small airway epithelial cells. Supports the growth of these cells over a period of at least 15 population doublings.