Learn More
Lifeline Cell Technology M MEL HU LIFELINE KT
SDP
Catalog No. 50311901
Encompass_Preferred
Includes DermaLife Basal Medium 485 mL, DermaLife M LifeFactors Kit. Low serum, PMA-free, Cholera toxin-free medium optimized for the culture of Human Epidermal Melanocytes over a period of at least 15 population doublings.
Catalog No. 50-311-901 Supplier Lifeline Cell Technology Supplier No. LL0027
Includes DermaLife Basal Medium 485 mL, DermaLife M LifeFactors Kit. Low serum, PMA-free, Cholera toxin-free medium optimized for the culture of Human Epidermal Melanocytes over a period of at least 15 population doublings.