Learn More
Cayman Chemical m-Methoxybenzamide, Purity: ≥98%, CAS: 5813-86-5, 25 g
SDP
Catalog No. 502988375
Encompass_Preferred
A PARP inhibitor (Ki = <2 µM); inhibits human placental ADP ribosyl transferase activity by 93% at 100 µM; increases the pain threshold in the paw pinch test in rats at 100, 200, and 300 mg/kg
Catalog No. 50-298-8375 Supplier Cayman Chemical Supplier No. 3045125 G
A PARP inhibitor (Ki = <2 µM); inhibits human placental ADP ribosyl transferase activity by 93% at 100 µM; increases the pain threshold in the paw pinch test in rats at 100, 200, and 300 mg/kg