Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-methyl red | 20691-84-3 | MFCD00059641 | >98.0% | 269.30 g/mol | C15H15N3O2 | 10 G
SDP

Catalog No. 5000283655
Encompass_Preferred

m-Methyl red is an organic pH indicator and biochemical reagent supplied as a solid for laboratory use. It is commonly used to monitor pH changes in buffer systems and biochemical assays where its color transition is useful for identifying acidic conditions. The substance has formula C15H15N3O2, molecular weight 269.30 g/mol, and CAS 20691-84-3.

  • pH indicator dye with color change in acidic range
  • Suitable for biochemical and microbiological assays
  • Solid powder form, typically supplied in 10 G packages
  • Chemical formula C15H15N3O2; molecular weight 269.30 g/mol
  • CAS number 20691-84-3 for unambiguous identification
  • SDS and certificate of analysis typically available

Catalog No. 50-002-83655 Supplier Medchemexpress LLC Supplier No. HYW13012810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-Methyl red is an organic pH indicator and biochemical reagent supplied as a solid for laboratory use. It is commonly used to monitor pH changes in buffer systems and biochemical assays where its color transition is useful for identifying acidic conditions. The substance has formula C15H15N3O2, molecular weight 269.30 g/mol, and CAS 20691-84-3.

  • pH indicator dye with color change in acidic range
  • Suitable for biochemical and microbiological assays
  • Solid powder form, typically supplied in 10 G packages
  • Chemical formula C15H15N3O2; molecular weight 269.30 g/mol
  • CAS number 20691-84-3 for unambiguous identification
  • SDS and certificate of analysis typically available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.