Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Methylanisole,MP Biomedicals

Catalog No. ICN213519 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213519 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213519 Supplier MP Biomedicals, Inc Supplier No. 0521351910

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100-84-5
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00008395
InChI Key OSIGJGFTADMDOB-UHFFFAOYSA-N
Synonym 3-methylanisole, 3-methoxytoluene, m-methylanisole, m-cresol methyl ether, m-methoxytoluene, benzene, 1-methoxy-3-methyl, anisole, m-methyl, methyl m-tolyl ether, m-cresyl methyl ether, methyl m-cresyl ether
PubChem CID 7530
IUPAC Name 1-methoxy-3-methylbenzene
SMILES COC1=CC=CC(C)=C1

Specifications

Quantity 10 g
Formula Weight 122.17
Chemical Name or Material M-Methylanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.