Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Methylbenzyl Alcohol,MP Biomedicals

Catalog No. ICN227322 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN227322 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN227322 Supplier MP Biomedicals, Inc Supplier No. 0522732205

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 587-03-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
InChI Key JJCKHVUTVOPLBV-UHFFFAOYSA-N
Synonym 3-methylbenzyl alcohol, 3-methylphenyl methanol, m-tolyl carbinol, m-tolylmethanol, benzenemethanol, 3-methyl, unii-ttg38p37u6, m-methylbenzyl alcohol, 3-methylbenzylalcohol, m-methyl benzyl alcohol, 3-methyl-benzenemethanol
PubChem CID 11476
ChEBI CHEBI:27995
IUPAC Name (3-methylphenyl)methanol
SMILES CC1=CC(=CC=C1)CO

Specifications

Quantity 10 g
Formula Weight 122.17
Chemical Name or Material M-Methylbenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.