Learn More
Lifeline Cell Technology M MICROV HU LIFELINE KT
SDP
Catalog No. 50311894
Encompass_Preferred
Includes VascuLife Basal Medium 475 mL, VascuLife VEGF-Mv LifeFactors (with Vascular Endothelial Growth Factor). Optimized for the culture of human microvascular endothelial cells in a 5% serum environment. No antimicrobials, no phenol red
Catalog No. 50-311-894 Supplier Lifeline Cell Technology Supplier No. LL0005
Includes VascuLife Basal Medium 475 mL, VascuLife VEGF-Mv LifeFactors (with Vascular Endothelial Growth Factor). Optimized for the culture of human microvascular endothelial cells in a 5% serum environment. No antimicrobials, no phenol red