Learn More
Lifeline Cell Technology M MICROV HU LIFELINE KT
SDP
Catalog No. 50311893
Encompass_Preferred
VascuLife Basal Medium 475 mL, VascuLife ENGS-Mv LifeFactors Kit (with Endothelial Cell Growth Supplement). Optimized for the culture of human microvascular endothelial cells in a 5% serum enviroment. No antimicrobials, no phenol red.
Catalog No. 50-311-893 Supplier Lifeline Cell Technology Supplier No. LL0004
VascuLife Basal Medium 475 mL, VascuLife ENGS-Mv LifeFactors Kit (with Endothelial Cell Growth Supplement). Optimized for the culture of human microvascular endothelial cells in a 5% serum enviroment. No antimicrobials, no phenol red.