Learn More
BIOHIPPO M-MLV, 1 mg
SDP
Catalog No. 5000856883
Encompass_Preferred
M-MLV Reverse Transcriptase synthesizes a complementary DNA strand initiating from a primer using either RNA (cDNA synthesis) or single-stranded DNA as a template. Recommended applications include cDNA Synthesis, Reverse Transcription (cDNA Synthesis), RT-PCR cDNA Synthesis, RT-qPCR, RT-PCR and cDNA Synthesis, PCR. Expressed in E.coli. Supplied as 1 mg.
Catalog No. 50-008-56883 Supplier BIOHIPPO Supplier No. BHP21409929
M-MLV Reverse Transcriptase synthesizes a complementary DNA strand initiating from a primer using either RNA (cDNA synthesis) or single-stranded DNA as a template. Recommended applications include cDNA Synthesis, Reverse Transcription (cDNA Synthesis), RT-PCR cDNA Synthesis, RT-qPCR, RT-PCR and cDNA Synthesis, PCR. Expressed in E.coli. Supplied as 1 mg.