Learn More
Lifeline Cell Technology M MSC HU LIFELINE KT
SDP
Catalog No. 50311904
Encompass_Preferred
Includes StemLife MSC Basal Medium 475 mL, StemLife MSC LifeFactors Kit. Optimized for the culture of human mesenchymal stem cells. Supports the undifferentiated expansion of mesenchymal stem cells. Contains no antimicrobials, no phenol red
Catalog No. 50-311-904 Supplier Lifeline Cell Technology Supplier No. LL0034
Includes StemLife MSC Basal Medium 475 mL, StemLife MSC LifeFactors Kit. Optimized for the culture of human mesenchymal stem cells. Supports the undifferentiated expansion of mesenchymal stem cells. Contains no antimicrobials, no phenol red