Learn More
New England Biolabs, Inc. M-MuLV Reverse Transcriptase – 50000 units
SDP
Catalog No. 50811670
Encompass
Moloney Murine Leukemia Virus (M-MuLV, MMLV) Reverse Transcriptase is an RNA-directed DNA polymerase. This enzyme can synthesize a complementary DNA strand initiating from a primer using either RNA (cDNA synthesis) or single-stranded DNA as a template. (1-4). M-MuLV Reverse Transcriptase lacks 3 5 exonuclease activity.
Catalog No. 50-811-670 Supplier New England Biolabs, Inc. Supplier No. M0253L
Moloney Murine Leukemia Virus (M-MuLV, MMLV) Reverse Transcriptase is an RNA-directed DNA polymerase. This enzyme can synthesize a complementary DNA strand initiating from a primer using either RNA (cDNA synthesis) or single-stranded DNA as a template. (1-4). M-MuLV Reverse Transcriptase lacks 3 5 exonuclease activity.