Learn More
New England Biolabs, Inc. ProtoScript II Reverse Transcriptase – 10000 units
SDP
Catalog No. NC0319405
Encompass
ProtoScript II Reverse Transcriptase is a recombinant M-MuLV reverse transcriptase with reduced RNase H activity and increased thermostability. It can be used to synthesize first strand cDNA at higher temperatures than the wild type M-MuLV. The enzyme is active up to 48C, providing higher specificity, higher yield of cDNA and more full-length cDNA product up to 12 kb.
Catalog No. NC0319405 Supplier New England Biolabs, Inc. Supplier No. M0368L
ProtoScript II Reverse Transcriptase is a recombinant M-MuLV reverse transcriptase with reduced RNase H activity and increased thermostability. It can be used to synthesize first strand cDNA at higher temperatures than the wild type M-MuLV. The enzyme is active up to 48C, providing higher specificity, higher yield of cDNA and more full-length cDNA product up to 12 kb.